Anti-SSRP1 Rabbit Monoclonal Antibody [clone: EPR7894]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129109-100EA 6580 DKK
ABCAAB129109-100
Anti-SSRP1 Rabbit Monoclonal Antibody [clone: EPR7894]
Antistoffer
Type: Primary
Antigen: SSRP1 (structure specific recognition protein 1)
Clonality: monoclonal
Clone: EPR7894
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu