Anti-ACO2 Rabbit Monoclonal Antibody [clone: EPR8283]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129105-100EA 6850 DKK
ABCAAB129105-100
Anti-ACO2 Rabbit Monoclonal Antibody [clone: EPR8283]
Antistoffer
Type: Primary
Antigen: ACO2 (aconitase 2, mitochondrial)
Clonality: monoclonal
Clone: EPR8283
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu