Anti-SERCA1 ATPase Rabbit Monoclonal Antibody [clone: EPR7322]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129104-100EA 3913.86 DKK
ABCAAB129104-100
Anti-SERCA1 ATPase Rabbit Monoclonal Antibody [clone: EPR7322]
Antistoffer
Type: Primary
Antigen: SERCA1 ATPase
Clonality: monoclonal
Clone: EPR7322
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR