Anti-ZNF136 Rabbit Monoclonal Antibody [clone: EPR8225]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129100-100EA 5840 DKK
ABCAAB129100-100
Anti-ZNF136 Rabbit Monoclonal Antibody [clone: EPR8225]
Antistoffer
Type: Primary
Antigen: ZNF136 (zinc finger protein 136)
Clonality: monoclonal
Clone: EPR8225
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR