Anti-SUOX Rabbit Monoclonal Antibody [clone: EPR7618]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129094-100EA 3354.74 DKK
ABCAAB129094-100
Anti-SUOX Rabbit Monoclonal Antibody [clone: EPR7618]
Antistoffer
Type: Primary
Antigen: SUOX (sulfite oxidase)
Clonality: monoclonal
Clone: EPR7618
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR