Anti-GRB7 Rabbit Monoclonal Antibody [clone: EPR6858]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129091-100EA 6000 DKK
ABCAAB129091-100
Anti-GRB7 Rabbit Monoclonal Antibody [clone: EPR6858]
Antistoffer
Type: Primary
Antigen: GRB7 (growth factor receptor-bound protein 7)
Clonality: monoclonal
Clone: EPR6858
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu