Anti-PF4 Rabbit Monoclonal Antibody [clone: EPR7762]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129090-100EA 4062.96 DKK
ABCAAB129090-100
Anti-PF4 Rabbit Monoclonal Antibody [clone: EPR7762]
Antistoffer
Type: Primary
Antigen: PF4 (platelet factor 4)
Clonality: monoclonal
Clone: EPR7762
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR