Anti-CDC27 Rabbit Monoclonal Antibody [clone: EPR7798]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129085-100EA 6000 DKK
ABCAAB129085-100
Anti-CDC27 Rabbit Monoclonal Antibody [clone: EPR7798]
Antistoffer
Type: Primary
Antigen: CDC27 (cell division cycle 27)
Clonality: monoclonal
Clone: EPR7798
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR