Anti-KARS Rabbit Monoclonal Antibody [clone: EPR7921]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129080-100EA 3764.76 DKK
ABCAAB129080-100
Anti-KARS Rabbit Monoclonal Antibody [clone: EPR7921]
Antistoffer
Type: Primary
Antigen: KARS (lysyl-tRNA synthetase)
Clonality: monoclonal
Clone: EPR7921
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR