Anti-USP24 Rabbit Monoclonal Antibody [clone: EPR7060]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129064-100EA 6010 DKK
ABCAAB129064-100
Anti-USP24 Rabbit Monoclonal Antibody [clone: EPR7060]
Antistoffer
Type: Primary
Antigen: USP24 (ubiquitin specific peptidase 24)
Clonality: monoclonal
Clone: EPR7060
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR