Anti-PIM2 Rabbit Monoclonal Antibody [clone: EPR6987]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129057-100EA 6630 DKK
ABCAAB129057-100
Anti-PIM2 Rabbit Monoclonal Antibody [clone: EPR6987]
Antistoffer
Type: Primary
Antigen: PIM2 (pim-2 oncogene)
Clonality: monoclonal
Clone: EPR6987
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR