Anti-BCAP29 Rabbit monoclonal antibody unconjugated [clone: EPR7781]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129023-100EA 5840 DKK
ABCAAB129023-100
Anti-BCAP29 Rabbit monoclonal antibody unconjugated [clone: EPR7781]
Antistoffer
Type: Primary
Antigen: BCAP29 (B-cell receptor-associated protein 29)
Clonality: monoclonal
Clone: EPR7781
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR