Anti-PMS1 Rabbit Monoclonal Antibody [clone: EPR8058]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129020-100EA 3692.6 DKK
ABCAAB129020-100
Anti-PMS1 Rabbit Monoclonal Antibody [clone: EPR8058]
Antistoffer
Type: Primary
Antigen: PMS1 (PMS1 postmeiotic segregation increased 1 (S. cerevisiae))
Clonality: monoclonal
Clone: EPR8058
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR