Anti-TCP1 epsilon Rabbit Monoclonal Antibody [clone: EPR7562]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129016-100EA 6030 DKK
ABCAAB129016-100
Anti-TCP1 epsilon Rabbit Monoclonal Antibody [clone: EPR7562]
Antistoffer
Type: Primary
Antigen: TCP1 epsilon
Clonality: monoclonal
Clone: EPR7562
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR