Anti-Exportin-5 Rabbit monoclonal antibody unconjugated [clone: EPR8452]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129006-100EA 6690 DKK
ABCAAB129006-100
Anti-Exportin-5 Rabbit monoclonal antibody unconjugated [clone: EPR8452]
Antistoffer
Type: Primary
Antigen: Exportin-5
Clonality: monoclonal
Clone: EPR8452
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu