Anti-INHBA Rabbit Monoclonal Antibody [clone: EPR2785]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB128958-100EA 6850 DKK
ABCAAB128958-100
Anti-INHBA Rabbit Monoclonal Antibody [clone: EPR2785]
Antistoffer
Type: Primary
Antigen: INHBA (inhibin, beta A)
Clonality: monoclonal
Clone: EPR2785
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR