Anti-RREB1 Mouse Monoclonal Antibody [clone: MM0485-5X25]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB90449-50EA 5070 DKK
ABCAAB90449-50
Anti-RREB1 Mouse Monoclonal Antibody [clone: MM0485-5X25]
Antistoffer
Type: Primary
Antigen: RREB1 (ras responsive element binding protein 1)
Clonality: monoclonal
Clone: MM0485-5X25
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human