Anti-AtRAC3 Rabbit polyclonal antibody unconjugated

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB50678-100EA 4900 DKK
ABCAAB50678-100
Anti-AtRAC3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: AtRAC3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR