Anti-PAX9 Rat Monoclonal Antibody [clone: 7C2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28538-100EA 5970 DKK
ABCAAB28538-100
Anti-PAX9 Rat Monoclonal Antibody [clone: 7C2]
Antistoffer
Type: Primary
Antigen: PAX9 (paired box 9)
Clonality: monoclonal
Clone: 7C2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR