Anti-Phosphotyrosine Mouse monoclonal antibody TRITC (Rhodamine) [clone: PY20]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28346-100EA 4270 DKK
ABCAAB28346-100
Anti-Phosphotyrosine Mouse monoclonal antibody TRITC (Rhodamine) [clone: PY20]
Type: Primary
Antigen: Phosphotyrosine
Clonality: monoclonal
Clone: PY20
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR