Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28335-50EA 2740 DKK
ABCAAB28335-50
Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]
Antistoffer
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEM-188
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR