Anti-ADAM23 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28302-100EA 4104.08 DKK
ABCAAB28302-100
Anti-ADAM23 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ADAM23 (ADAM metallopeptidase domain 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR