Anti-BUB1B Sheep Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28194-125EA 5500 DKK
ABCAAB28194-125
Anti-BUB1B Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR