Anti-XIAP Mouse Monoclonal Antibody [clone: 2F1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28151-100EA 5620 DKK
ABCAAB28151-100
Anti-XIAP Mouse Monoclonal Antibody [clone: 2F1]
Antistoffer
Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: monoclonal
Clone: 2F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR