Anti-CYC Mouse monoclonal antibody [clone: 7H8.2C12]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28137-100EA 4060 DKK
ABCAAB28137-100
Anti-CYC Mouse monoclonal antibody [clone: 7H8.2C12]
Type: Primary
Antigen: CYC (cytochrome C)
Clonality: monoclonal
Clone: 7H8.2C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR