Anti-CASP7 Mouse monoclonal antibody [clone: 10-1-62]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28130-100EA 4990 DKK
ABCAAB28130-100
Anti-CASP7 Mouse monoclonal antibody [clone: 10-1-62]
Type: Primary
Antigen: CASP7 (caspase 7, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 22920
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR