Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: LT8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB28010-50EA 2850 DKK
ABCAAB28010-50
Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: LT8]
Antistoffer
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone: LT8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR