Anti-ITGA9/B1 Mouse Monoclonal Antibody [clone: Y9A2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27947-100EA 4098.45 DKK
ABCAAB27947-100
Anti-ITGA9/B1 Mouse Monoclonal Antibody [clone: Y9A2]
Antistoffer
Type: Primary
Antigen: ITGA9/B1 (integrin, alpha 9/beta 1)
Clonality: monoclonal
Clone: Y9A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR