Anti-TIE1 Mouse Monoclonal Antibody [clone: Cl. 8C9]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27851-100EA 5670 DKK
ABCAAB27851-100
Anti-TIE1 Mouse Monoclonal Antibody [clone: Cl. 8C9]
Antistoffer
Type: Primary
Antigen: TIE1 (tyrosine kinase with immunoglobulin-like and EGF-like domains 1)
Clonality: monoclonal
Clone: Cl. 8C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR