Anti-WNT2 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27794-100EA 5880 DKK
ABCAAB27794-100
Anti-WNT2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: WNT2 (wingless-type MMTV integration site family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR