Anti-BCL3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27780-100EA 5670 DKK
ABCAAB27780-100
Anti-BCL3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BCL3 (B-cell CLL/lymphoma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR