Anti-V5 Tag Mouse Monoclonal Antibody (Biotin) [clone: SV5-Pk1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27672-50EA 5630 DKK
ABCAAB27672-50
Anti-V5 Tag Mouse Monoclonal Antibody (Biotin) [clone: SV5-Pk1]
Antistoffer
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: monoclonal
Clone: SV5-Pk1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR