Anti-DDX4 / MVH Mouse monoclonal antibody unconjugated [clone: mAbcam27591]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27591-100EA 5960 DKK
ABCAAB27591-100
Anti-DDX4 / MVH Mouse monoclonal antibody unconjugated [clone: mAbcam27591]
Antistoffer
Type: Primary
Antigen: DDX4 / MVH
Clonality: monoclonal
Clone: mAbcam27591
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR