Anti-BMP7 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27569-100EA 5620 DKK
ABCAAB27569-100
Anti-BMP7 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BMP7 (bone morphogenetic protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR