Anti-DDIT3 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27539-100EA 4360 DKK
ABCAAB27539-100
Anti-DDIT3 Rabbit polyclonal antibody
Type: Primary
Antigen: DDIT3 (DNA-damage-inducible transcript 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR