Anti-ADAP1 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27476-100EA 4098.45 DKK
ABCAAB27476-100
Anti-ADAP1 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ADAP1 (ArfGAP with dual PH domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR