Anti-IL6ST Mouse Monoclonal Antibody [clone: B-K11]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27399-100EA 5670 DKK
ABCAAB27399-100
Anti-IL6ST Mouse Monoclonal Antibody [clone: B-K11]
Antistoffer
Type: Primary
Antigen: IL6ST (interleukin 6 signal transducer (gp130, oncostatin M receptor))
Clonality: monoclonal
Clone: B-K11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR