Anti-A2M Mouse Monoclonal Antibody [clone: HYB339-02]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27161-100EA 5330 DKK
ABCAAB27161-100
Anti-A2M Mouse Monoclonal Antibody [clone: HYB339-02]
Antistoffer
Type: Primary
Antigen: A2M (alpha-2-macroglobulin)
Clonality: monoclonal
Clone: HYB339-02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR