Anti-S-Mecoprop Mouse monoclonal antibody [clone: HYB337-01]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB27153-100EA 4250 DKK
ABCAAB27153-100
Anti-S-Mecoprop Mouse monoclonal antibody [clone: HYB337-01]
Type: Primary
Antigen: S-Mecoprop
Clonality: monoclonal
Clone: HYB337-01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR