Anti-6X His tag Rabbit Polyclonal Antibody (Biotin)
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB27025-100EA 4402.56 DKK
ABCAAB27025-100
Anti-6X His tag Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: 6X His tag (6X His tag)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu