Anti-SOX9 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26414-100EA 3357.88 DKK
ABCAAB26414-100
Anti-SOX9 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SOX9 (SRY (sex determining region Y)-box 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR