Anti-MAFA Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB26405-100EA 3651.34 DKK
ABCAAB26405-100
Anti-MAFA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAFA (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human