Anti-Exendin 4 Mouse monoclonal antibody unconjugated [clone: 35]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26263-100EA 5330 DKK
ABCAAB26263-100
Anti-Exendin 4 Mouse monoclonal antibody unconjugated [clone: 35]
Antistoffer
Type: Primary
Antigen: Exendin 4
Clonality: monoclonal
Clone: 35
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR