Anti-HA tag Rabbit Polyclonal Antibody (Biotin)

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26228-250EA 5630 DKK
ABCAAB26228-250
Anti-HA tag Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR