Anti-NEUROG2 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26190-100EA 4460 DKK
ABCAAB26190-100
Anti-NEUROG2 Rabbit polyclonal antibody
Type: Primary
Antigen: NEUROG2 (neurogenin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR