Anti-CARD14 Chicken polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26165-50EA 4140 DKK
ABCAAB26165-50
Anti-CARD14 Chicken polyclonal antibody
Type: Primary
Antigen: CARD14 (caspase recruitment domain family, member 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR