Anti-PAG1 Mouse monoclonal antibody Dyomics® 647 [clone: MEM-255]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26145-100EA 4060 DKK
ABCAAB26145-100
Anti-PAG1 Mouse monoclonal antibody Dyomics® 647 [clone: MEM-255]
Type: Primary
Antigen: PAG1 (phosphoprotein associated with glycosphingolipid microdomains 1)
Clonality: monoclonal
Clone: MEM-255
Conjugation: Dyomics® 647
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR