Anti-BCL2L1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7B2.5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB26035-50EA 5330 DKK
ABCAAB26035-50
Anti-BCL2L1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7B2.5]
Antistoffer
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: monoclonal
Clone: 7B2.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR