Anti-MPO Mouse Monoclonal Antibody [clone: 2C7]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB25989-100EA 4359.26 DKK
ABCAAB25989-100
Anti-MPO Mouse Monoclonal Antibody [clone: 2C7]
Antistoffer
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone: 2C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human