Anti-TNFSF18 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25948-100EA 5670 DKK
ABCAAB25948-100
Anti-TNFSF18 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TNFSF18 (tumor necrosis factor (ligand) superfamily, member 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR